missing translation for 'onlineSavingsMsg'
Learn More

Ilaprazole, MedChemExpress

Product Code. 16416217
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16416217

Brand: Medchem Express HY101664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ilaprazole (IY-81149) is an orally active proton pump inhibitor. Ilaprazole irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of pump inhibitory activity of 6 μM in rabbit parietal cell preparation. Ilaprazole is used for the research of gastric ulcers. Ilaprazole is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.

Ilaprazole (IY-81149) is an orally active proton pump inhibitor. Ilaprazole irreversibly inhibits H+/K+-ATPase in a dose-dependent manner with an IC50 of pump inhibitory activity of 6 μM in rabbit parietal cell preparation. Ilaprazole is used for the research of gastric ulcers. Ilaprazole is also a potent TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor.

Specifications

Chemical Name or Material Ilaprazole
CAS 172152-36-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C19H18N4O2S
Quantity 5 mg
Synonym IY-81149
Solubility Information DMSO : ≥ 35 mg/mL (95.51 mM) ∣Ethanol : 12.5 mg/mL (34.11 mM; Need ultrasonic)
SMILES O=S(C1=NC2=CC=C(N3C=CC=C3)C=C2N1)CC4=NC=CC(OC)=C4C
Molecular Weight (g/mol) 366.44
Formula Weight 366.44
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.