Learn More
Cytiva ExoProStar™ 1-step
Use for enzymatic PCR clean-up. Cytiva Lifescience™ Illustra ExoProStar™ 1-step with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs.
£85.75 - £2770.00
Specifications
Description | Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube. |
---|---|
Isolation Technology | Enzymatic PCR Clean-up Technology |
For Use With (Application) | Remove unincorporated primers and dNTPs |
Product Type | illustra ExoProStar 1-Step |
Quantity | 1 Each |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12124082
|
Cytiva
US77702 |
100 |
£85.75
100 reactions |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12134082
|
Cytiva
US77720 |
2000 |
£1232.00
2000 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11941411
|
Cytiva
US77705 |
500 |
£339.00
500 reactions |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11961411
|
Cytiva
US77750 |
5000 |
£2770.00
5000 reactions |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- One tube with one mix of enzymes delivering PCR and sequence reaction clean-up
- Just one pipetting step to set up your reaction
- Fast 30-minute, hands-free protocol
- 100% recovery of PCR product
- Scalable for different reaction sizes
- Automation friendly
Specifications
Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase combined in one tube to remove unincorporated primers and dNTPs. For maximum convenience comes in a single tube. | |
Remove unincorporated primers and dNTPs | |
1 Each |
Enzymatic PCR Clean-up Technology | |
illustra ExoProStar 1-Step | |
100% recovery of PCR product |