missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
£268.00 - £1440.36
Specifications
Description | Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. |
---|---|
For Use With (Application) | PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product |
Product Type | illustra ExoProStar S |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15239764
|
Cytiva
US79050 |
500 |
£268.00
500 reactions |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15229764
|
Cytiva
US79200 |
2000 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15249764
|
Cytiva
US79500 |
5000 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Easy to automate
- Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
- Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
- Scalable for different reaction sizes
- No loss of PCR product
- Complete heat inactivation of the enzymes within 10 minutes
- Fast 15 minute protocol
- Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
- Improves digestion efficiency with no degradation of the target PCR product
- Sample is completely intact and ready for immediate use in manual or automated processes
Specifications
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. | |
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product | |
illustra ExoProStar S |