Learn More
Cytiva GenomiPhi V2 DNA Amplification Kit
Amplify whole genomes with high efficiency and representation. Cytiva Life Sciences™ illustra™ GenomiPhi V2 DNA Amplification Kit provides whole genome amplification with yields of 4 to 7μg.
Brand: Cytiva 25-6600-31
410.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GE
Additional Details : CAS Number : 7732-18-5 Weight : 0.02610kg
Description
- Starting material can be purified DNA or nonpurified cell lysates
- Simple protocol provides typical yields of 4 to 7μg DNA and average product lengths >10kb
- Less hands-on time compared to traditional isolation methods
- Representative amplification of the whole genome
- Automation-friendly protocol with great reproducibility
- High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
- Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification
Specifications
Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C. Thaw components on ice and maintain at 0°C to 4°C during handling. | |
Genomic DNA Amplification | |
100 | |
> 10 kb |
Illustra GenomiPhi V2 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 4 to 7 μg yield from nanogram amounts of DNA sample. | |
0.9mL Sample Buffer, 0.9mL Reaction Buffer, 100μL Enzyme mix, 20μL Control DNA (lambda) | |
illustra GenomiPhi V2 DNA Amplification Kit | |
1.5-2 h |
Safety and Handling
- GenomiPhi v2 DNA Amplification Kit, 100 reactions
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-1.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 1.5% of components with unknown hazards to the aquatic environment.