Learn More
Cytiva GFX 96 PCR Purification Kit
Simultaneously purify up to 96 PCR products of between 100 bp and 10 kbp in as little as 15 minutes
Brand: Cytiva 28-9034-45
1280.77 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GH
Additional Details : CAS Number : 593-84-0 Weight : 1.20000kg
Includes: Binding plates; Wash plates; Collection plates and color-coded bottles of capture buffer; Wash buffer; Choice of two elution buffers (Tris-HCl and sterile water); Instruction booklet
Description
- High yields of pure DNA recovered in a small volume of water or a low-ionic strength buffer
- Typical recoveries are > 85% for PCR products of 100 bp to 10kb length; Salt removal > 99%
- Prepacked glass-fiber matrix 96-well plates are validated for use with the Macherey Nagel NucleoVac 96-vacuum manifold
- No need for ethanol precipitations and hazardous organic extractions
Fluorescent sequencing, microarrays, labeling, hybridization, ligation, and transformation
Specifications
Kit uses glass fiber matrix technology in a 96-well format for highly efficient purification of PCR products. DNA fragments from PCR are captured by the matrix in presence of a chaotropic salt and contaminants are removed by washing matrix. | |
Fluorescent sequencing, microarrays, labeling, hybridization, ligation and transformation | |
illustra GFX 96 PCR Purification Kit | |
100 bp-10 kbp |
DNA Solutions, Enzyme Reactions, PCR Mixtures | |
Glass-fiber matrix 96-well plates binding plates, wash plates, collection plates and color-coded bottles of capture Buffer, Wash Buffer choice of two Elution Buffers (Tris HCl and sterile Water), instruction booklet | |
96-Well Plates | |
Up to 95% when purifying a 910 bp fragment at a starting concentration of 8.4 ng/μl |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .