missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GFX™ PCR DNA and Gel Band Purification Kit

Product Code. p-3676775 Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
No. of Reactions:
100 Preps
250 Preps
missing translation for 'unitSize'
100 reactions
250 reactions
This item is not returnable. View return policy

Product Code. 10536295

missing translation for 'mfr': Cytiva 28903470

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

GFX columns; Collection tubes; Color-coded bottles of capture buffer; Wash buffer; two elution buffers: Tris-HCl and sterile water

Purify and concentrate PCR products or DNA fragments ranging in size from 50 bp to 10 kb. Cytiva Life Sciences™ illustra™ GFX™ PCR DNA and Gel Band Purification Kit can be used to purify DNA from reaction volumes up to 100μL or agarose gel slices of up to 900mg.

Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution.

  • Rapid and easy-to-use purification method with less than 10 minutes hands-on time
  • Flexible 10 to 50μL elution volume for different DNA concentration needs
  • Improved column design with increased maximum volume holding of up to 800μL liquid volume (950μL without a cap)
  • DNA of exceptional purity (to 99.5%) is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning
  • Visual color indicator added to the capture buffer to ensure optimal pH for maximum DNA binding and recovery
  • Kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin-column for the purification of DNA from both solution and agarose gel

TRUSTED_SUSTAINABILITY

Specifications

Description Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg.
Sample Type Agarose Gel Slices, DNA Solutions, Enzyme Reactions, PCR Mixtures
Formulation Tris HCl and sterile Water
For Use With (Application) Isolation and concentration of DNA fragments (50bp to 10kb) from PCR mixtures, DNA-containing Agarose Gel bands, Enzyme-based DNA modifications and restriction digestions
Includes GFX™ columns, collection tubes, color-coded bottles of capture Buffer, Wash Buffer, two Elution Buffers
No. of Reactions 100 Preps
Product Type illustra GFX PCR DNA and Gel Band Purification Kit
Sample Volume 50 bp-10 kb
Yield PCR 10 μl elution volume-65%, PCR 50 μl elution volume-82%, 300 mg agarose 10 μl elution volume-57%, 300 mg agarose 50 μl elution volume-91%
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : thiocyanic acid salts
  • UNKNOWN ACUTE-5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 5% of components with unknown hazards to the aquatic environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.