Learn More
Cytiva MicroSpin™ S-200 HR Columns
For rapid purification of labeled single-stranded or double-stranded DNA fragments ≥50 bases in length in a volume of 25 to 100μL using spin-column chromatography
Brand:  Cytiva 27-5120-01
224.20 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 11713309
  • £241.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Prepacked with Sephacryl™ S-200 HR medium pre-equilibrated in TE buffer
  • Percent sample recovery: >80%
  • Tested in nickase, single- and double-stranded exonuclease and RNase assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spin Column
Rapid purification of labeled single-stranded or double-stranded DNA fragments <50 bases in length in a volume of 25 to 100μL using spin-column chromatography
Column