Learn More
Cytiva MicroSpin™ S-400 HR Columns
Use for rapid purification of PCR products (>200 bp) from unincorporated primers (<32-mers) and nucleotides. Cytiva Life Sciences™ illustra™ MicroSpin™ S-400 HR Columns use spin-column chromatography for quick results.
Brand: Cytiva 27-5140-01
227.40 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.18000kg
Description
- Accommodates 25 to 50μL for post-PCR cleanup prior to cloning or a second amplification reaction; 25 to 100μL for all other applications
- Ready to use—requires <4 min. from sample application to collection of purified product
- Prepacked with Sephacryl™ S-400 HR media pre-equilibrated in TE buffer
- Percent sample recovery: >80%
- Tested in nickase, single- and double-stranded exonuclease and RNase assays
Specifications
Spin Column | |
Rapid purification of PCR products (>200bp) from unincorporated primers (<32-mers) and Nucleotides using spin-column chromatography | |
Column | |
Each |