missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NAP™ Columns, NAP-25

Use for rapid and efficient purification of DNA and oligonucleotides (>10-mer) in less than 15 minutes by gravity flow. Cytiva Life Sciences™ illustra™ NAP™ Columns are disposable columns prepacked with Sephadex™ G-25 DNA Grade

Brand:  Cytiva 17-0852-02

448.61 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : GH

Additional Details : CAS Number : 9048-46-8 Weight : 1.00620kg


 View more versions of this product

Product Code. 10186464

  • £487.00 / Pack of 50
In Stock
to see stock.

Description

Description

  • Disposable columns prepacked with Sephadex™ G-25 DNA Grade Gel require only gravity to run
  • Available in three sizes depending on sample volume: 0.5mL (NAP-5), 1mL (NAP-10) or 2.5mL (NAP-25)
  • Useful for small-scale purification, desalting, and buffer exchange

Specifications

Specifications

Rapid and efficient purification of DNA and Oligonucleotides
Complete step-by-step protocol identifying the fixed volumes needed for equilibration, washing and sample elution
NAP-25
50 Pack
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : reaction mass of:5-chloro2-methyl-4-isothiazolin-3-one¢EC no. 247-500-7! and 2-methyl-2H-isothiazol-

SDS
Documents

Documents

Certificates
Special Offers

Special Offers