missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NICK™ Columns

Product Code. 10597065 Shop All Cytiva Products
Click to view available options
Quantity:
20 Columns
50 Columns
Unit Size:
Pack of 20
Pack of 50
This item is not returnable. View return policy

Product Code. 10597065

Brand: Cytiva 17085501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For rapid purification of labeled DNA (>20 bases in length) from unincorporated radiolabeled nucleotides using gravity-flow chromatography

  • Prepacked with Sephadex™ G-50 DNA Grade in distilled water with 0.15% Kathon™ CG/ICP Biocide
  • Typical recovery: ≥90% of applied DNA
  • Can accommodate sample volumes up to 100μL

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Rapid purification of labeled DNA from unincorporated radiolabeled Nucleotides using gravity-flow chromatography
Includes Sephadex™ G-50 DNA grade in distilled Water with 0.15% Kathon™ CG/ICP Biocide
Product Type Column
Quantity 20 Columns
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : reaction mass of: 5-chloro2-methyl-4-isothiazolin-3- one ¢ECno. 247-500-7! and2-methyl-2H-isothiaz
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.