missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva plasmidPrep Mini Spin Kit
For the rapid isolation of high and low copy number plasmid DNA
£107.05 - £338.00
Specifications
Description | Purify high yields of plasmid DNA in less than 10 min. Color-coded tubes and columns simplify the protocol and purified DNA is suitable for all common downstream applications. |
---|---|
For Use With (Application) | Suitable for many downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCRSequencing, PCR, ligation and cloning, restriction digest, enzymatic modification, template for labeled probe generation, transforma |
Includes | Spin columns prepacked with a novel Silica membrane, Suspension, Lysis and Neutralization Buffers containing chaotropic salts, Wash and Elution Buffers, microcentrifuge collection tubes, full protocol booklet, Detachable, quick reference protocol card |
Product Type | illustra plasmidPrep Mini Spin Kit |
Purity | 1.7 |
Includes: Spin columns prepacked with a novel silica-membrane; Suspension, lysis, and neutralization buffers containing chaotropic salts; Wash and elution buffers; Microcentrifuge collection tubes; Full protocol booklet; Detachable, quick reference protocol card
Product Code | Brand | No. of Reactions | Sample Volume | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Sample Volume | Price | Quantity & Availability | ||||
10379434
|
Cytiva
28-9042-70 |
250 Preps | 1.5 mL |
£338.00
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10772047
|
Cytiva
28-9042-69 |
50 Preps | 3.0 mL |
£107.05
50 reactions |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The illustra plasmidPrep Mini Spin Kit uses a simple plasmid DNA purification protocol involving a modified alkaline lysis procedure and a novel silica-based membrane to achieve highly efficient plasmid DNA purification. Instead of organic solvents, the kit contains chaotropic salts to denature protein components and promote selective binding of DNA to the novel silica membrane.
- Total preparation time of less than 10 minutes—representing a 50% reduction in protocol time
- Produces high-quality plasmid DNA with excellent reproducibility for use in cloning, restriction enzyme digestion, PCR amplification, and DNA sequencing
- Increased ease of use provided minimal changes in pipetting volume from one part of the protocol to the next, less centrifugation steps, color-coded kit components, and lack of organic solvents
- The typical plasmid yields from a fresh 1.5 ml culture of E.coli containing a high copy number plasmid is approximately 9 μg
- Can accommodate high and low copy number plasmid purification
Specifications
Purify high yields of plasmid DNA in less than 10 min. Color-coded tubes and columns simplify the protocol and purified DNA is suitable for all common downstream applications. | |
Spin columns prepacked with a novel Silica membrane, Suspension, Lysis and Neutralization Buffers containing chaotropic salts, Wash and Elution Buffers, microcentrifuge collection tubes, full protocol booklet, Detachable, quick reference protocol card | |
1.7 | |
6-15 μg |
Suitable for many downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCRSequencing, PCR, ligation and cloning, restriction digest, enzymatic modification, template for labeled probe generation, transforma | |
illustra plasmidPrep Mini Spin Kit | |
Approx. 9 min |