Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Pre-mixed, pre-dispensed reaction beads optimized for performing standard PCR amplifications. Cytiva Lifescience™ illustra™ PuReTaq Ready-To-Go™ PCR Beads are stable at ambient temperature.
Brand: Cytiva 27-9559-01
This item is not returnable.
View return policy
Description
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
Specifications
Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer. | |
1 Kit | |
96 |
0.2mL Tube with hinged cap | |
Standard PCR | |
200μL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Product Title
Cytiva PuReTaq Ready-To-Go™ PCR Beads > 0.2mL hinged tube w/cap; Pack size: 96 reactions
Spot an opportunity for improvement?Share a Content Correction