Learn More
Cytiva Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit
Highly efficient and representative whole-genome amplification with 40 to 60μg yield from nanogram amounts of DNA sample. Cytiva Lifescience™ illustra™ Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit is part of the Phi29 DNA polymerase family of products.
Brand:  Cytiva 25-6601-96
521.55 GBP valid until 2025-12-31
Use promo code "24339" to get your promotional price.
 View more versions of this product
Product Code. 12967160
  • £549.00 / Each
Explore more special offers
Description
Description
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
  • The reaction mixture is provided as predispensed, single-dose, lyophilized cakes in either strips of 8 tubes, or 96-well plates.
  • Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample.
  • Amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.
  • The average product length is over 10kb.
  • Procedure is quick, simple and does not require a thermocycler.
  • A typical DNA yield of 12 to 20μg DNA can be achieved in less than two hours from only 10ng of genomic DNA input.
  • The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or a nonpurified cell lysate may be used.
  • Protocols for the amplification of DNA from blood lysates and dried blood spots are included in the manual.
  • GenomiPhi amplified DNA is suitable for various applications such as genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For whole genome amplification by isothermal strand displacement amplification; typical downstream applications are genotyping (SNP, STR, array CGH) and DNA archiving.
Ready-To-Go GenomiPhi V3 cakes, 1 × 96-well plate; 2x Denaturation buffer, 1 × 1.2 ml; Control DNA (Lambda), 500 ng; 12 Strips of 8 domed caps
96
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-72.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 72.5% of components with unknown hazards to the aquatic environment.
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit > 96 Reactions