missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ready-To-Go™ RT-PCR Beads
Optimized for performing either one-step or conventional two-step reverse transcription. Cytiva Lifescience™ Illustra™ Ready-To-Go™ RT-PCR Beads are ambient temperature stable, pre-mixed, and ready for pre-dispensed reactions.
£366.00 - £408.00
Product Code | Brand | Format | No. of Reactions | Volume | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | No. of Reactions | Volume | Price | Quantity & Availability | |||
10187044
|
Cytiva
27-9259-01 |
0.2mL Tube with hinged cap | 96 | 200μL |
£366.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10064974
|
Cytiva
27-9267-01 |
0.2mL Tube | 96 | 200μL |
£379.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10015264
|
Cytiva
27-9266-01 |
0.5mL Tube | 100 | 500μL |
£408.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Reagents are optimized for full-length cDNA synthesis (>7.5 kb) and optimal sensitivity from PCR
- Provided in either thin walled 0.5 ml or 0.2 ml tubes/plates compatible with most thermocyclers
- Supplied with both pd(N)6 and oligo(dT) primers for flexibility
- The Ready-To-Go manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
Specifications
1 Kit | |
RT-PCR |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.