missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Single Cell GenomiPhi™ DNA Amplification Kit

Amplify whole genome DNA from as little as a single cell. Cytiva Lifescience™ illustra™ Single Cell GenomiPhi™ DNA Amplification Kit combines unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.

Brand:  Cytiva 29108039

 View more versions of this product

Product Code. 15311652

  • £1443.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Single Cell GenomiPhi Reaction buffer 1 x 275µL, Single Cell GenomiPhi Enzyme mix 1 x 25µL, Single Cell GenomiPhi Amplification mix 1 x 25µL, Single Cell GenomiPhi DTT Solution 1 x 100µL, Single Cell GenomiPhi Lysis buffer 1 x 100µL and Single Cell GenomiPhi Neutralisation buffer 1 x 100µL
This item is not returnable. View return policy

Description

Description

  • For whole genome amplification of genomic DNA from as little as a single cell
  • Novel clean-up process supresses background allowing sensitivity down to 1fg of input DNA
  • Simple Isothernal strand displacement (MDA) technology using Phi29 DNA polymerase
  • Amplifications can be completed in less than 3 hours with less than 15 minutes hands-on time
  • Optimized to promote complete cell lysis with uniform denaturation of DNA prior to amplification
  • Reaction time: 2 to 4 hours
  • Product average size: >10kb
Applications

For SNP arrays, Next Gen Sequencing, aCGH

Recommended Storage

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C


TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.
illustra™ Single Cell GenomiPhi™ DNA Amplification Kit
Single Cell GenomiPhi Reaction buffer 1 x 275μL, Single Cell GenomiPhi Enzyme mix 1 x 25μL, Single Cell GenomiPhi Amplification mix 1 x 25μL, Single Cell GenomiPhi DTT Solution 1 x 100μL, Single Cell GenomiPhi Lysis buffer 1 x 100μL and Single Cell GenomiPhi Neutralisation buffer 1 x 100μL
illustra Single Cell GenomiPhi DNA Amplification Kit
2 to 4 hours
Single Cell GenomiPhi DNA Amplification kit combines the unique capabilities of Phi29 DNA polymerase with an optimized formulation to amplify genomic DNA from 1-1000 cells without background interference.
Genomic DNA Amplification
100
>10kb
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.