missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTP Sets

Product Code. p-4907751 Shop All Cytiva Products
Click to view available options
Quantity:
4 x 25 μm
4 x 100 μm
4 x 500 μm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 10084724

Brand: Cytiva 28406551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use dNTP sets for optimum performance and consistency. Cytiva Lifescience™ illustra™ Solution dNTP Sets are free from DNase, RNase, and nicking enzyme activity.

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7
Product Type dNTP set
Concentration 100 mM
Purity >99%
Quantity 4 x 25 μm
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Identifier
  • dATP, 100MMOLAR SOLN, 25 UMOL; part of 'dNTP Set, 100 mM Solutions (dATP, dCTP, dGTP, dTTP), 4 x 25
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.