Learn More
Cytiva Solution dNTPs
Use dNTPs incorporating the most recent advances in manufacturing technology. Cytiva illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
Brand:  Cytiva 28-4065-32
 View more versions of this product
Product Code. 10527065
  • £125.63 / Each
Explore more special offers
Description
Description
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
dTTP
>99%
Colorless
7
100 mM
100 μm
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling