missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva triplePrep Kit
Shop All Cytiva Products
Click to view available options
:
TriplePrep Kit; 50 preps
Description
Includes
Protocol and a supply of components sufficient for 50 preps.
- High-yield process allows extraction/isolation from single limited-size samples from biopsies, archived tissues, and tumors
- Isolated products suitable for downstream applications such as PCR, restriction digestion, sequencing, array CGH, RT-qPCR, gene expression microarray, SDS-PAGE, Western blotting, 2-D DIGE, and LCMS
- Flexible workflow allows easy isolation of any two or all three analytes
- Color-coded caps and bottles match to protocol steps to minimize the chance for error
Specifications
Specifications
| Description | The illustra triplePrep Kit is designed for rapid, simultaneous extraction and isolation of high quality genomic DNA (gDNA), total RNA, and total denatured proteins from animal tissues and mammalian cells. |
| For Use With (Application) | Extraction and isolation of high quality genomic DNA, total RNA and total denatured Proteins from animal tissues and mammalian cells |
| Includes | Protocol and a supply of components sufficient for 50 preps |
| No. of Reactions | 50 Preps |
| Product Type | illustra triplePrep kit |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : TCA (ISO)
- UNKNOWN ACUTE-27.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction