missing translation for 'onlineSavingsMsg'
Learn More

Cytiva IMAC Sepharose™ High Performance Media

For optimizing purification of histidine-tagged proteins using Immobilized Metal Affinity Chromatography (IMAC) when Ni2+ is not the best choice of metal ion

Brand:  Cytiva 17-0920-07


Product Code. 11534955

This item is not returnable. View return policy

Description

Description

  • 34μm beads of highly cross-linked 6% agarose, to which a chelating group has been covalently coupled
  • Charge with transition metal of choice for optimized selectivity
  • Small bead size for high performance purification
  • High binding capacity: ≥40mg histidine-tagged protein/mL medium when charged with Ni2+
  • Allows more sample to be applied; a more concentrated target protein is obtained more at a lower cost/mg and in less time
  • Stable in pH range 3-12 (long term), and in all commonly used buffers

Avoid use with chelating agents, e.g., EDTA, EGTA, or citrate.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

<300 cm/hr.
34 μm
4°C to 30°C
Purification of Polyhistidine tagged Proteins
3 to 12
IMAC Sepharose HP
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • IMAC sepharose high perfomance

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers