missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent

Dramatically reduces background fluorescence resulting from non-specific staining during secondary detection using conjugates of streptavidin or secondary antibodies

Brand:  Molecular Probes™ R37107

160.67 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.03000kg

Product Code. 13552550

  • £241.00 / Each
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent dramatically reduces background fluorescence resulting from non-specific staining during secondary detection using conjugates of streptavidin or secondary antibodies. This reagent is applied directly to slides or coverslips containing fixed and permeabilized cell or tissue samples prior to staining with fluorescent probes.
  • Easy protocol—apply ready-made solution using a dropper bottle
  • Compatible with the other steps in your immunostaining protocol
  • Effective with most fluorescent labels
  • Stable at room temperature—keep handy at your scope or cell culture area

Cell imaging applications
Image-iT™ FX Signal Enhancer reagent is a highly effective product for blocking background staining that results from the nonspecific interactions of a wide variety of fluorescent dyes with cell and tissue constituents. Background staining seen with fluorescent conjugates of streptavidin and antibody conjugates are largely eliminated when the Image-iT™ FX Signal Enhancer reagent is applied to fixed and permeabilized cells prior to staining.

Suggestions for use
  • Apply Image-iT™ FX Signal Enhancer reagent directly to slides or coverslips containing cell or tissue samples prior to immune-staining.
  • After the fix/perm step, simply wash and add enough drops to cover your sample, then incubate for 30 min and proceed with your normal staining protocol.
  • Additional blocking steps may be performed subsequent to blocking with the Image-iT™ Signal Enhancer ReadyProbes™ Reagent, as needed

Recommended Storage

Store at ≤ 25°C

Specifications

Specifications

Signal Enhancers
Cellular Imaging
6 × 2.5mL dropper bottles
1 kit
Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent Kit, Colorless Solution, pH 7, Fluorescent Detection Method, Signal Enhancers Reagent, Contains 6 x 2.5mL Dropper Bottles, Used for Cellular Imaging, ≤ 25°C Storage Temperature
Fluorescent
≤25°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers