missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Image-iT™ Green Hypoxia Reagent

Product Code. 15834154
Click to view available options
Quantity:
1 Vial
5 Vials
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 15834154

Brand: Invitrogen™ I14834

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells.

Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.

Features of Image-iT Green Hypoxia Reagent include:
• Indicates hypoxia in live cells by fluorescing in low oxygen environments
• Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
• Easy to use—just add to cell culture media and image

Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.

Order Info

  • Store at -5 to -30°C. Protect from light.
  • Stable for at least 6 months after receiving.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -30°C. Protect from light.
  • Stable for at least 6 months after receiving.
  • Product Type Reagent
    Product Line Image-iT
    Quantity 1 Vial
    Reagent Type Hypoxia Reagent
    Shipping Condition Approved for shipment on Wet or Dry Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.