missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant
Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.
£138.00 - £235.00
Specifications
Content And Storage | Store at 15°C to 25°C. Do not freeze. When stored as directed, product is stable for (at least 6 months after receipt. ) |
---|---|
Product Line | Image-iT™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
16329583
|
Invitrogen™
V11331 |
30 mL |
£138.00
30mL |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
16319583
|
Invitrogen™
V11330 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.
• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415
Specifications
Store at 15°C to 25°C. Do not freeze. When stored as directed, product is stable for (at least 6 months after receipt. ) |
|
Image-iT™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.