Learn More
Invitrogen™ Image-iT™ TMRM Reagent (mitochondrial membrane potential indicator)
Healthy mitochondrial membranes maintain a difference in electrical potential between the interior and exterior of the organelle, referred to as a membrane potential.
Brand: Invitrogen™ I34361
152.76 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Tetramethylrhodamine, methyl ester (TMRM) is a cell-permeant dye that accumulates in active mitochondria with intact membrane potentials. If the cells are healthy and have functioning mitochondria, the signal will be bright. Upon loss of the mitochondrial membrane potential, TMRM accumulation will cease and the signal will dim or disappear. TMRM signal can be detected with fluorescence microscopy, flow cytometry, cell sorting, high-throughput screening, and high content analysis.
• For live-cell dynamic study of mitochondrial membrane potential (not fixable)
• Detectable with RFP/TRITC filter set (absorbance peak: 548 nm, emission peak: 574 nm)
• multiplexable with green, deep red, violet, and blue emission dyes, such as GFP, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
Specifications
Orange | |
Reagent | |
5 x 100 μL |
Store in freezer -5°C to -30°C and protect from light. | |
Molecular Probes™ | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.