missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ IMAGEN™ Adenovirus Kit using Direct Immunofluorescence Assay Product Code.: 13215779

Thermo Scientific™ IMAGEN™ Adenovirus Kit using Direct Immunofluorescence Assay

Product Code. 13215779
50 Tests, 50 tests
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
This item is not returnable. View return policy

Product Code. 13215779

Brand: Thermo Scientific™ K6100112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly detect adenovirus antigen in clinical samples and confirm adenovirus in cell culture with this direct immunofluorescence assay.

Rapidly detect adenovirus antigen in clinical samples and confirm adenovirus in cell cultures with Thermo Scientific™ IMAGEN™ Adenovirus Kit. This direct immunofluorescence assay utilizes a genus-specific monoclonal antibody to detect an epitope of adenovirus hexon proteins, which is expressed in all known human adenovirus serotypes1.

Human adenoviruses are associated with a wide range of clinical diseases in both immunocompetent and immunocompromised individuals, which include infections of the respiratory tract, conjunctiva and gastrointestinal tract2,3. Infections are common in children and can occur sporadically or in outbreaks.

Use IMAGEN Adenovirus Kit as a rapid, sensitive and specific method for direct detection and identification of human adenovirus serotypes in clinical specimens or cell cultures.

  • Rapid results—within little over 30 minutes of receiving test sample or culture
  • Simple—ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity—high quality, well characterized antibodies used in the assay provide excellent sensitivity and specificity
  • Easy-to-read results
  • Long shelf life—24 months

Adenoviruses are non-enveloped DNA viruses of icosahedral symmetry. The family Adenoviridae comprises two genera, mammalian adenoviruses (Mastadenoviruses) and avian adenoviruses (Aviadenoviruses)4. At least 47 known serotypes of human adenovirus have been identified and characterized by hemagglutination, neutralization, DNA-hybridization and restriction endonuclease analysis of adenoviral DNA2,4,5.

The laboratory diagnosis of adenovirus infection plays an important role in patient management and enables effective management and control of outbreaks. A range of techniques have been used to confirm the identification of adenovirus isolates, including neutralization tests, radioimmunoassay, DNA hybridization, electron microscopy and DNA electrophoretyping6,7,8,9,10. These techniques can be complex, laborious and often inappropriate for routine use. Recently indirect immunofluorescence tests or enzyme immunoassays (e.g., IDEIA™ PCE Chlamydia Kit) using genus-specific monoclonal or polyclonal antibodies have been described for the direct detection of adenovirus in clinical specimens or cell culture monolayers10,11.

Direct immunofluorescence tests utilizing specific monoclonal antibodies offer a rapid, sensitive and specific method for the direct detection of adenoviruses in clinical specimens such as nasopharyngeal aspirates and conjunctival smears or for the confirmation of adenovirus isolates in cell culture monolayers. IMAGEN Adenovirus Kit is a direct immunofluorescence test for the detection and identification of human adenovirus serotypes in clinical specimens or cell cultures.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description IMAGEN™ Adenovirus Kit
Detectable Analytes Adenovirus antigen
Includes Positive control slide, mounting fluid, adenovirus reagent, instructions for use
Quantity 50 Tests

Instructions

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number6245571 Certificate TypeCertificate of Analysis Date04/08/2025 Product Code
Lot Number6159677 Certificate TypeCertificate of Analysis Date12/12/2024 Product CodeK610011-2
Lot Number6127320 Certificate TypeCertificate of Analysis Date19/07/2024 Product CodeK610011-2
Lot Number3794233 Certificate TypeCertificate of Analysis Date09/05/2024 Product CodeK610011-2
Lot Number3766578 Certificate TypeCertificate of Analysis Date15/02/2024 Product CodeK610011-2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IMAGEN™ Adenovirus Kit using Direct Immunofluorescence Assay >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.