missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IMAGEN™ hMPV Positive Control Slide

Validate your test procedures with hMPV Control Slide.

Brand:  Thermo Scientific™ S612830-2

Product Code. 13225329

  • £669.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Validate your test procedures with Thermo Scientific™ IMAGEN™ hMPV Positive Control Slide, a Teflon-coated glass microscope slide with two separate 6mm diameter well areas containing acetone-fixed human fibroblast cells infected with hMPV antigen.

Sealed in foil pouch filled with nitrogen.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IMAGEN hMPV Control Slide
1 Slide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6212356 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog NumberS612830-2
Lot Number6202358 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberS612830-2
Lot Number3619079 Certificate TypeCertificate of Analysis Date09/03/2023 Catalog NumberS612830-2
Lot Number3350942 Certificate TypeCertificate of Analysis Date03/09/2021 Catalog NumberS612830-2
Lot Number3247331 Certificate TypeCertificate of Analysis Date08/04/2021 Catalog NumberS612830-2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IMAGEN™ hMPV Positive Control Slide >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.