missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IMAGEN™ Human Metapneumovirus (hMPV) Kit using Direct Immunofluorescence Assay

Carry out rapid detection of hMPV in clinical samples with this direct immunofluorescence assay.

Brand:  Thermo Scientific™ K612511-2

453.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 13225789

  • £494.00 / 50 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rapidly detect hMPV in clinical samples with Thermo Scientific™ IMAGEN™ Human Metapneumovirus (hMPV) Kit. This direct immunofluorescence assay utilizes two monoclonal antibodies in order to detect specific structural proteins expressed in all strains of human metapneumovirus.

Human metapneumovirus (hMPV) is a respiratory virus that causes an upper respiratory infection. It causes seasonal epidemics centred around the winter months, and symptoms include fever with upper respiratory symptoms such as cough and nasal congestion1, 2. However, hMPV has also been associated with more severe illness and lower respiratory tract symptoms such as pneumonia and bronchiolitis3,4.

Use IMAGEN Human metapneumovirus (hMPV) Kit as a rapid, sensitive and specific method for direct detection of hMPV in clinical samples such as nasopharyngeal aspirates.

  • Rapid results- within little over 30 minutes of receiving test sample or culture
  • Simple- ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to-read results
  • Long Shelf Life- typically 24 months.

Human metapneumovirus (hMPV) is an enveloped, spherical RNA virus of the family Paramyxoviridae, subfamily Paramyxovirinae and is classified within the genus Metapneumovirus3. hMPV is the only member of the genus that affects humans, with the other members including avian pneumoviruses A, B, C and D1,5.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
Human metapneumovirus (hMPV) antigen
50 Tests
IMAGEN Human metapneumovirus Kit
2 x 1 positive control slide, mounting fluid, hMPV Reagent, instruction for use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6127663 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberK612511-2
Lot Number3712441 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog NumberK612511-2
Lot Number3640650 Certificate TypeCertificate of Analysis Date29/06/2023 Catalog NumberK612511-2
Lot Number3581516 Certificate TypeCertificate of Analysis Date25/02/2023 Catalog NumberK612511-2
Lot Number3562941 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog NumberK612511-2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IMAGEN™ Human Metapneumovirus (hMPV) Kit using Direct Immunofluorescence Assay >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.