missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IMAGEN™ Respiratory Syncytial Virus Kit (RSV) using Direct Immunofluorescence Assay

Carry out rapid detection of Respiratory Syncytical Virus (RSV) in clinical samples with this direct immunoflurescence assay.

Brand:  Thermo Scientific™ K610211-2

470.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 13235779

  • £490.00 / 50 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Respiratory Syncytial Virus (RSV) Antigen
Reagent
50 Tests
IMAGE Respiratory Syncytial Virus (RSV) Kit
Nasopharyngeal
2 x positive control slide, mounting fluid, RSV test reagent, instruction for use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Instructions

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6135730 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog NumberK610211-2
Lot Number3773424 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberK610211-2
Lot Number3660978 Certificate TypeCertificate of Analysis Date08/09/2023 Catalog NumberK610211-2
Lot Number3611148 Certificate TypeCertificate of Analysis Date19/04/2023 Catalog NumberK610211-2
Lot Number3434607 Certificate TypeCertificate of Analysis Date30/08/2022 Catalog NumberK610211-2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IMAGEN™ Respiratory Syncytial Virus Kit (RSV) using Direct Immunofluorescence Assay >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.