Learn More
Thermo Scientific™ IMAGEN™ Herpes Simplex Virus (HSV) Kit using Direct Immunofluorescence Assay
Carry out rapid detection and identification of HSV1 and HSV2 in cell cultures with this direct immunofluorescence assay.
Brand:  Thermo Scientific™ K610611-2
478.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13275779
  • £511.00 / 50 tests
Explore more special offers
Description
Description
The IMAGEN™ Herpes Simplex Virus typing test is a qualitative direct immunofluorescence test for the detection and typing of HSV 1 and HSV 2 in cell cultures.
HSV is a common and universal infection of humans, associated with a wide range of clinical diseases in both immunocompetent and immunocompromised individuals. Both HSV 1 and 2 are frequently implicated in localised vesicular infections of the skin, conjunctiva and mucous membranes of the mouth or genitalia1, 2,3.
Use IMAGEN Herpes Simplex Virus (HSV) Kit as a rapid, sensitive and specific method for the detection and differentiation of HSV1 and HSV2 isolates in cell culture monolayers.
  • Rapid results- within little over 30 minutes of receiving test sample or culture
  • Simple- ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity- high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity.
  • Easy-to-read results
  • Long Shelf Life- typically 24 months.
Herpes simplex virus is a DNA virus containing an icosahedral nucleocapsid surrounded by a lipid containing envelope. Human HSV is classified within the family Herpesviridae and is a member of the sub-family Alphaherpesvirinae. The genus Simplexvirus includes two type species of human herpes simplex virus, HSV 1 and HSV 24.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
HSV glycoproteins (HSV-1/2)
50 Tests
IMAGEN Herpes Simplex Virus (HSV) Kit
2 x 2 positive control slide, mounting fluid, HSV type 1 reagent, HSV type 2 reagent, instruction for use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ IMAGEN™ Herpes Simplex Virus (HSV) Kit using Direct Immunofluorescence Assay >