missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMC-1C11 Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541798
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
IMC-1C11 is an anti-angiogenesis agent. It is a chimeric anti-kinase insert domain-containing receptor (KDR) antibody that blocks VEGFR-KDR interaction and inhibits VEGFR-induced endothelial cell proliferation. IMC-1C11 is used for treatment of patients with liver metastases from colorectal carcinoma. IMC-1C11 is a chimerized antibody that targets the KDR receptor (also referred to as VEGFr) on vascular endothelial cells by inhibiting binding of the essential ligand, vascular endothelial growth factor (VEGF), to its receptor. KDR is a key receptor associated with tumor angiogenesis. As solid tumors cannot grow efficiently without new blood supply, use of IMC-1C11 results in the inhibition of tumor growth and death of tumor cells by apoptosis.
Spezifikation
IMC-1C11 | |
Recombinant Monoclonal | |
Unconjugated | |
IMC-1C11; VEGFA | |
Human VEGFA | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, SDS-Page | |
1.2 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
Chemical |
Produktvorschläge
Customers who viewed this item also viewed
Viewing 1-5 of 12
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
IMC-1C11 Recombinant Human Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur