missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IMDM
IMDM
£13.59 - £122.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Concentration | 1 X |
Form | Liquid |
Product Type | IMDM Medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11510596
|
Gibco™
21980032 |
500 mL |
£13.59
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11520596
|
Gibco™
21980065 |
10 x 500 mL |
£122.00
5000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ IMDM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ IMDM product made in our Grand Island facility (12440-053). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
1 X | |
IMDM Medium | |
No α±-Thioglycerol, No 2-Mercaptoethanol | |
6.9 to 7.3 |
2 to 8°C | |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture | |
For in-vitro diagnostic use |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title