missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IMDM, GlutaMAX™ Supplement
IMDM, GlutaMAX™ Supplement
Brand: Gibco™ 31980048
203.36 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
Iscove's Modified Dulbecco's Medium (IMDM) is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells. We offer a variety of Gibco™ IMDM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins. In addition, this unique medium lacks iron, with potassium nitrate replacing ferric nitrate. Gibco™ IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ IMDM is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ IMDM product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
IMDM contains no proteins, lipids, or growth factors. Therefore, IMDM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). IMDM uses a sodium bicarbonate buffer system (3.024 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Iscove's Modified Dulbecco's Medium, 10 x 500mL, Liquid, 1 X Concentration, 6.9 to 7.3 pH, With Glutamax™, Phenol Red, Sodium Pyruvate, 12 Months Shelf Life, 270 to 310mOsm/kg Osmolality, For Mammalian Cell Culture | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture | |
10 x 500 mL |
2 to 8°C | |
1 X | |
Iscove's modified Dulbecco's medium | |
No α±-Thioglycerol, No 2-Mercaptoethanol | |
6.9 to 7.3 |