missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imetit dihydrobromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
100 mg
Description
An extremely potent, high affinity agonist at H3 and H4 receptors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 50 mg |
Solubility Information | Freely soluble in water,practically insoluble in acetone /n |
Formula Weight | 332.06 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Imetit dihydrobromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'