Learn More
Imibenconazole solution, Honeywell Fluka™
100 μg/mL in acetonitrile, analytical standard, reference material
Brand: Honeywell Fluka R1356-2ML
Additional Details : CAS Number : 75-05-8 Weight : 0.00100kg Transport : UN number : 1648 Chem class : 3 Pack group : II
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Liquid | |
Analytical Standard | |
82°C | |
6°C |
86598-92-7,75-05-8 | |
Glass Bottle | |
2 mL | |
Fungicides and Bactericides |