missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, Thermo Scientific™

Product Code. 10269562
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 132213-07-1
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
InChI Key IEOADZOTWIMSMC-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-6-ylmethanol, 6-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-methanol, imidazo 1,2-a pyridin-6-yl-methanol, imidazo 1,2-a pyridin-6-yl methanol, acmc-209zmz, imidazo 1,2-a pyridin-6yl methanol, 6-hydroxymethyl imidazo 1,2-a pyridin, 4-hydroimidazo 1,2-a pyridin-6-ylmethan-1-ol
PubChem CID 7060530
IUPAC Name imidazo[1,2-a]pyridin-6-ylmethanol
SMILES C1=CC2=NC=CN2C=C1CO

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridin-6-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.