missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10187692
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6200-60-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD01720458
InChI Key DOGXPDFZEQXZDS-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylicacid, pubchem19678, wln: t56 an dnj bvq, 3-carboxyimidazo 1,2-a pyridine, h-imidazo 1,2-a pyridine-3-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, 1h-imidazo 1,2-a pyridine-3-carboxylic acid
PubChem CID 73142
IUPAC Name imidazo[1,2-a]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=C2C=CC=CN12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.