missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 11353297
Click to view available options
Quantity:
5 g
25 g
10111-08-7
C4H4N2O
96.09
MFCD00003544
XYHKNCXZYYTLRG-UHFFFAOYSA-N
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
24955
1H-imidazole-2-carbaldehyde
O=CC1=NC=CN1
10111-08-7
C4H4N2O
96.09
MFCD00003544
XYHKNCXZYYTLRG-UHFFFAOYSA-N
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole
24955
1H-imidazole-2-carbaldehyde
O=CC1=NC=CN1

Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole-2-carboxaldehyde is a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with an important application to treat type-2 diabetes. It is used in the preparation of tridentate Schiff-base carboxylate-containing ligands by undergoing condensation reaction with amino acids beta-alanine and 2-aminobenzoic acid. It is also involved in the study of the imidazole-directed allylation of aldimines.

Solubility
Soluble in water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∽205°C (decomposition)
Quantity 25 g
Beilstein 4371302
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 96.09
Percent Purity 97%
Chemical Name or Material Imidazole-2-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'