missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
£78.40 - £254.00
Chemical Identifiers
CAS | 10111-08-7 |
---|---|
Molecular Formula | C4H4N2O |
Molecular Weight (g/mol) | 96.09 |
MDL Number | MFCD00003544 |
InChI Key | XYHKNCXZYYTLRG-UHFFFAOYSA-N |
Synonym | imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole |
PubChem CID | 24955 |
IUPAC Name | 1H-imidazole-2-carbaldehyde |
SMILES | O=CC1=NC=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10577624
|
Thermo Scientific Acros
398470010 |
1 g |
£78.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10745125
|
Thermo Scientific Acros
398470050 |
5 g |
£254.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10111-08-7 | |
96.09 | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
24955 | |
O=CC1=NC=CN1 |
C4H4N2O | |
MFCD00003544 | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
1H-imidazole-2-carbaldehyde |
Specifications
10111-08-7 | |
80°C (0.1 mmHg) | |
Glass Bottle | |
MFCD00003544 | |
XYHKNCXZYYTLRG-UHFFFAOYSA-N | |
1H-imidazole-2-carbaldehyde | |
24955 | |
98% |
209°C | |
97.5% min. (HPLC) | |
C4H4N2O | |
imidazole-2-carboxaldehyde, 2-formylimidazole, 1h-imidazole-2-carboxaldehyde, imidazole-2-carbaldehyde, 2-imidazolecarboxaldehyde, imidazole-2-aldehyde, 1h-imidazolecarboxaldehyde, 2-formyl imidazole, imidazol-2-carbaldehyde, 2-formyl-1h-imidazole | |
O=CC1=NC=CN1 | |
96.09 | |
96.09 | |
Imidazole-2-carboxaldehyde |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Imidazole-2-carboxaldehyde
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only