missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imipenem monohydrate, 98%

Product Code. p-7077543 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15505146

Brand: Thermo Scientific Chemicals 455322500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Imipenem monohydrate,Tienam, Primaxin, CAS # 74431-23-5 is a broad-spectrum carbapenem antibiotic that interferes with the activity of penicillin binding proteins along the inner cell wall.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Imipenem monohydrate is a beta-lactam derived from the compound thienamycin, produced by Streptomyces cattleya
• Disrupts cell wall integrity, preventing cell division and promoting lysis by interfering with the activity of penicillin binding proteins
• Imipenem is a third-generation cephalosporin that can kill both gram-negative and gram-positive bacteria

Applications
• Suitable for the study of antibiotic resistance
• High-quality, laboratory grade for use in vitro and in vivo experiments

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74431-23-5
Molecular Formula C12H19N3O5S
Molecular Weight (g/mol) 317.36
MDL Number MFCD09753321
InChI Key GSOSVVULSKVSLQ-XMCAPODCSA-N
PubChem CID 5282372
ChEBI CHEBI:51799
IUPAC Name (5R,6S)-3-[2-(aminomethylideneamino)ethylsulfanyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
SMILES O.C[C@H](O)[C@@H]1[C@H]2CC(SCCN=CN)=C(N2C1=O)C(O)=O

Specifications

Type Imipenem Monohydrate
Melting Point 193.0°C to 198.0°C
Infrared Spectrum Authentic
Water 5 to 8%
Quantity 250 mg
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material Imipenem monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.