missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Maleimide PEGylated mcKLH

Perform hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens with this sulfhydryl-reactive mariculture KLH.

Brand:  Thermo Scientific™ 77663

318.68 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 11800094

  • £478.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Upon receipt store product at -20°C. Product is shipped with an ice pack.
Imject Maleimide Activated Pegylated Mariculture KLH
10 mg
Imject™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZF400998 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number77663
Lot NumberYL383037 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77663
Lot NumberYG372953 Certificate TypeCertificate of Analysis Date04/08/2023 Catalog Number77663
Lot NumberYC366701 Certificate TypeCertificate of Analysis Date15/03/2023 Catalog Number77663
Lot NumberXI365343 Certificate TypeCertificate of Analysis Date06/10/2022 Catalog Number77663
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ Maleimide PEGylated mcKLH > Activated Pegylated mcKLH; 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.