Learn More
Thermo Scientific™ Imject™ Maleimide-Activated mcKLH Spin Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Imject Maleimide-Activated mcKLH is sulfhydryl-reactive immunogenic mariculture keyhole limpet hemocyanin (KLH) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
Highlights:
Convenient packages – choose stabilized, lyophilized Maleimide-Activated mcKLH or a complete immunogen conjugation kit
Optimized activation – specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of KLH, ensuring consistently high levels of conjugation
Sulfhydryl-reactive – engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure – complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein – KLH is isolated from the mollusk Megathura crenulata (keyhole limpet); mcKLH is a high-quality form of KLH that is harvested from limpets grown in mariculture
Highly immunogenic – KLH has a high molecular mass (0.5 to 13 million daltons; aggregates of 350 and 390kDa subunits) and elicits a stronger immune response than BSA or ovalbumin
Excellent solubility – our preparations of mariculture KLH are more soluble than typical forms and we also offer a PEGylated form for even greater hydrophilicity and solubility
Includes:
Kit contains stabilized, lyophilized Maleimide-Activated mcKLH (in PBS with stabilizer), buffers and desalting columns to easily prepare ready-to-use immunogens for immunization
Specifications
Specifications
| Content And Storage | Sufficient For: 5 conjugation reactions and complete immunogen preparations, each with up to 2 mg sulfhydryl-containing peptide • Imject Maleimide-Activated mcKLH, 5 x 2 mg • Imject Maleimide Conjugation Buffer, 30 mL • Imject Purification Buffer Salts, 5 x 0.84 g • Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns Store the Maleimide-Activated blue Carrier Protein at -20°C. Store remaining components at 4°C. |
| Description | Imject Maleimide Activated mcKLH Spin Kit |
| Format | Kit |
| Quantity | 5 x 2 mg |
| Product Line | Imject |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.