missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Imject™ Ovalbumin (in PBS)

Prepare hapten-carrier conjugates to generate antibodies using this conveniently formulated, purified ovalbumin (OVA) protein.

Brand:  Thermo Scientific™ 77120

Product Code. 11884061

  • £246.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Imject™ Ovalbumin Carrier Protein is purified ovalbumin protein conveniently formulated for preparation of hapten-carrier conjugates.

Because ovalbumin is only weakly immunogenic, its primary use in antibody production workflows is as a secondary (independent) carrier protein to make antigens more amenable to antibody screening procedures. For example, peptide antigens are more easily coated in polystyrene microplates when conjugated to ovalbumin, thereby facilitating the screening of antisera that were produced using immunogen conjugates with KLH or other more immunogenic carrier protein.

Highlights:

  • Purified from chicken egg whites and lyophilized in phosphate buffered saline
  • Single polypeptide protein with molecular mass of 45,000 daltons
  • Contains 20 lysine residues, most of which have primary amines that are capable of reacting with a conjugation reagent
  • Contains 14 aspartic acid and 33 glutamic acid residues whose carboxyl groups account for the net negative charge (pI 4.63) of ovalbumin
  • Not as immunogenic as keyhole limpet hemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent
  • Useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Supplied: Carrier proteins are supplied lyophilized. When reconstituted with 2mL of ultrapure water the buffer compositions for BSA and mcKLH is 0.1M sodium phosphate, 0.15M NaCl; pH 7.2 and for OVA is 0.01M sodium phosphate, pH 7.2. Proteins contain proprietary stabilizers.

Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature
5 x 20 mg
Imject™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ404878 Certificate TypeCertificate of Analysis Date26/10/2024 Catalog Number77120
Lot NumberZH402419 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number77120
Lot NumberZG401692 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number77120
Lot NumberZD398145A Certificate TypeCertificate of Analysis Date15/05/2024 Catalog Number77120
Lot NumberZC85003 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog Number77120
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Imject™ Ovalbumin (in PBS) > Ovalbumin in phosphate buffer; 5 x 20mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.