Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11766188
  • £157.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Designed for the Fisherbrand Dual Frequency Ultrasonic Cleaner models
  • Fine mesh bottom keeps small parts secure
  • Two handles for ease-of-use
  • Clean and simple design fits perfectly
  • Top-quality durable stainless steel
Specifications
Specifications
Basket
Ultrasonic Cleaner, 6.9 L
Stainless Steel