Learn More
Fisherbrand™ Immersion Oil
Use this immersion oil with biological compound microscopes. Fisherbrand™ Immersion Oil is available in 5mL bottles.
Brand: Fisherbrand™
Code : 46
Additional Details : CAS Number : 8012-95-1 Weight : 0.01080kg
Description
- Immersion Oil, 5mL
- Accessory for microscopes: FB69019 and FB69020
Specifications
Immersion Oil | |
5 mL |
Safety and Handling
- Immersion oil fluorescence-free "dropper-bottle"
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : paraffin oils