Learn More
Motic™ Immersion Oil
Transparent oil of high refractive index for use with oil immersion objectives.
Brand:  Motic™ 1101001300011
Product Code. 15485092
  • £16.40 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High refractive index.
  • Transparent.
  • Increases the numerical aperture of the objective lens.
  • Increases resolving power of a microscope.
Specifications
Specifications
Oil immersion objectives
5 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • cargille immersion oil
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : paraffins (petroleum), hydrogenated terphenyls, butene, terphenyl