Learn More
Cytiva Immobiline™ DryStrip Gels
Represent a comprehensive selection of narrow-, medium- and wide-range IPG strips. Cytiva Life Sciences™ Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application.
£101.90 - £496.00
Specifications
Description | Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. |
---|---|
Color | Colorless |
Product Type | Precast Gel |
Separation Type | Isoelectric Focusing |
Form | Dried |
Product Code | Brand | pH Range | Gradient | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Gradient | Length (Metric) | Price | Quantity & Availability | |||
11544985
|
Cytiva
17-1235-01 |
3 to 10 | Non-linear | 18 cm |
£148.46
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10753407
|
Cytiva
17-6001-97 |
6 to 11 | Linear | 18 cm |
£228.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11763239
|
Cytiva
17-6001-96 |
6 to 11 | Linear | 13 cm |
£421.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11743239
|
Cytiva
17-6001-94 |
6 to 11 | Linear | 7 cm |
£226.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11733249
|
Cytiva
17-6002-47 |
6 to 9 | Linear | 24 cm |
£496.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11713249
|
Cytiva
17-6002-45 |
3 to 10 | Non-linear | 24 cm |
£184.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10146414
|
Cytiva
17-6003-73 |
3 to 11 | Non-linear | 7 cm |
£167.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11793289
|
Cytiva
17-6003-75 |
3 to 11 | Non-linear | 13 cm |
£207.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11713299
|
Cytiva
17-6003-77 |
3 to 11 | Non-linear | 24 cm |
£185.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11524985
|
Cytiva
17-1233-01 |
4 to 7 | Linear | 18 cm |
£151.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11534985
|
Cytiva
17-1234-01 |
3 to 10 | Linear | 18 cm |
£148.47
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10545925
|
Cytiva
17-6001-11 |
3 to 10 | Linear | 7 cm |
£101.90
Pack of 12 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||
10732637
|
Cytiva
17-6001-12 |
3 to 10 | Non-linear | 7 cm |
£101.90
Pack of 12 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
11773229
|
Cytiva
17-6001-10 |
4 to 7 | Linear | 7 cm |
£101.90
Pack of 12 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
11703239
|
Cytiva
17-6001-13 |
4 to 7 | Linear | 13 cm |
£145.95
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11713239
|
Cytiva
17-6001-14 |
3 to 10 | Linear | 13 cm |
£145.95
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11723239
|
Cytiva
17-6001-15 |
3 to 10 | Non-linear | 13 cm |
£145.95
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11753239
|
Cytiva
17-6001-95 |
6 to 11 | Linear | 11 cm |
£450.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11723249
|
Cytiva
17-6002-46 |
4 to 7 | Linear | 24 cm |
£185.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10697465
|
Cytiva
17-6002-44 |
3 to 10 | Linear | 24 cm |
£184.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11703299
|
Cytiva
17-6003-76 |
3 to 11 | Non-linear | 18 cm |
£155.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10584375
|
Cytiva
17-6003-74 |
3 to 11 | Non-linear | 11 cm |
£169.00
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11723299
|
Cytiva
18-1016-60 |
4 to 7 | Linear | 11 cm |
£125.91
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
11733299
|
Cytiva
18-1016-61 |
3 to 10 | Linear | 11 cm |
£125.91
Pack of 12 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
Description
Choose from a comprehensive range of overlapping Immobiline DryStrip gels that cover narrow (1 pH unit), medium (3 to 5 pH units), and broad (8 pH units) pH ranges. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the proteins evenly over the gel length to obtain maximum resolution.
Five narrow pH range (1 pH unit), 24-cm IPG strips deliver high resolution and protein loading capacity, which results in well separated spots with high protein content after the second-dimension separation
- Select the broad pH 3 to 11 Non-Linear (NL) gradient Immobiline DryStrip gel for fast and efficient screening to gain a broad overview of total protein disribution
- For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range
- Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline DryStrip gels
- Use matching IPG Buffers for optimized running conditions
- Use IPG strips directly after reswelling without further manipulation
- Bar coded strips for convenient sample and batch tracking
- Immobiline DryStrip gels are rehydrated with a matching IPG Buffer prior to use
- Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein
- Samples can also be loaded after rehydration
Specifications
Immobiline DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. | |
Precast Gel | |
Dried | |
−20°C | |
0.05 cm | |
0.3 cm |
Colorless | |
Isoelectric Focusing | |
Industrial applications: analytical chemistry, research | |
0.19 in. | |
0.118 in. |