Learn More
Cytiva Immobiline™ DryStrip Kit and Components
Precast Immobiline DryStrip gels for running the first dimension of a 2-D electrophoresis experiment with the Multiphor II electrophoresis system
Brand: Cytiva 18-1004-35
This item is not returnable.
View return policy
Description
Run up to 12 Immobiline DryStrip strips simultaneously with up to 100μL on each Immobiline DryStrip.
- The instruction manual presents all necessary information for the entire method for horizontal 2-D electrophoresis. By appropriate adaptations, the SDS electrophoresis can also be performed using vertical electrophoresis equipment
- Immobiline DryStrip kit facilitates sample application and running and equilibration of Immobiline DryStrip for the first dimension in 2-D (two-dimensional) electrophoresis
- Isoelectric focusing with an immobilized pH gradient (IPG), makes true isoelectric focusing possible and significantly improves the reproducibility of the spot distribution along the pH gradient axis of 2-D maps
- Individual components are also available separately
Specifications
Multiphor II Electrophoresis System | |
Sample Cup |