Learn More
Cytiva Immobiline™ Sample Application Strips
Offer improved gel performance with faster and more stable gradients, fewer artifacts, and easier zymogram assays
Brand: Cytiva 18-1002-74
Code : GE
Additional Details : Weight : 0.04300kg
Description
- For use with SDS and immobiline samples
- Silicone applicator strip with sample holes to load samples onto Isoelectric focussing (IEF) Gels
Specifications
26 SDS sample | |
SDS Sample Application Strip | |
5 |