missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Immune Response Panel, 80plex

The ProcartaPlex Human Immune Response Panel 80plex enables the study of immune function by analyzing 80 protein targets in a single well using Luminex xMAP technology.

Brand:  Invitrogen™ EPX800-10080-901

8047.43 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 17807260

  • £8690.00 / Pack of 96

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The ProcartaPlex Human Immune Response Panel 80plex enables the study of immune function by analyzing 80 protein targets in a single well using Luminex xMAP technology. Analyze 80 cytokine, chemokine, and growth factor targets simultaneously for efficient immune response profiling, biomarker discovery, and validation. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.This kit is not compatible with the MAGPIX instrument.

Target [bead region]

  • Cytokines: G-CSF [42], GM-CSF [44], IFN alpha [48], IFN gamma [43], IL-10 [28], IL-12p70 [34], IL-13 [35], IL-15 [65], IL-16 [70], IL-17A [36], IL-18 [66], IL-1 alpha [62], IL-1 beta [18], IL-2 [19], IL-20 [81], IL-21 [72], IL-22 [76], IL-23 [63], IL-27 [14], IL-3 [73], IL-31 [37], IL-34 [79], IL-37 [7], IL-4 [20], IL-5 [21], IL-6 [25], IL-7 [26], IL-8 [27], IL-9 [52], LIF [15], M-CSF [67], MIF [53], TNF beta [54], TNF alpha [45], TSLP [80]
  • Chemokines: BLC [29], CCL1 (I-309) [83], CCL17 (TARC) [91], CCL21 (6Ckine/SLC) [96], CCL23 (MPIF) [98], CCL25 (TECK) [94], CXCL6 (GCP-2) [99], ENA-78 [82], Eotaxin [33], Eotaxin-2 [30], Eotaxin-3 [49], Fractalkine (CX3CL1) [59], Gro-alpha (CXCL1) [61], IP-10 (CXCL10) [22], I-TAC (CXCL11) [57], MCP-1 (CCL2) [51], MCP-2 (CCL8) [8], MCP-3 (CCL7) [68], MCP-4 (CCL13) [90], MDC (CCL22) [87], MIG (CXCL9) [69], MIP-1 alpha (CCL3) [12], MIP-1 beta (CCL4) [47], MIP-2 alpha (CXCL2) [95], MIP-3 beta (CCL19) [93], MIP-3 alpha (CCL20) [56]
  • Growth factors: NGF beta [55], FGF-2 [75], HGF [46], MMP-1 [64], SCF [39], VEGF-A [78]
  • Soluble receptors: APRIL [88], BAFF [86], CD30 [84], CD40L (CD154) [74], IL-2R (CD25) [9], TNF-RII [85], TRAIL [58], TREM-1 [13], TWEAK [97]
  • Serine proteases: Granzyme A [38], Granzyme B [77]
  • Lectin pentraxin: Gal-3 [89], PTX3 [100]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—the most referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

QuantiGene RNA testing—one machine, two applications

Gain deeper insight with complementary and highly correlative gene expression and protein assays. With the largest multiplex panel for measurement of protein and RNA, you can now profile more biomarkers with less sample. The Immune Response 80-Plex Human QuantiGene Plex Panel (Cat. No. QGP-180-10080) enables efficient immune response profiling and biomarker discovery.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multiplex Panel
Human
See Certificate of Analysis
Multiplex Kit
Less than 15%
ProcartaPlex™
Serum, Plasma: 25 μL, CCS: 50 μL
Wet Ice
96 Tests
Non-Combinable
See Certificate of Analysis
Luminex™ Instruments
Less than 15%
Serum, Plasma, Cell Culture Supernatants

• 4 vials Human Standard Mix (lyophilized)
• 2 vials Control High
• 2 vials Control Low
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (1X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Streptavidin-PE (1X)
• 8-Tube Strip
• Adhesive Film
• Flat-Bottom 96-well Plate, black

Store at 2°C to 8°C.

Immunology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.