Learn More
Thermo Scientific™ Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.
Brand: Thermo Scientific™ 95029180
255.60 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 2.52000kg
Description
Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.
Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.- For solid phase immuno assays, e.g. ELISA
- Choice of four surfaces for optimal binding
- Free-standing modules are easy to handle
- Alphanumeric system for easy orientation
- Fit standard equipment
- High optical quality for reliable results
- Polystyrene
Choice of Surface
- PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
- Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
- MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
- MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins
Specifications
Hydrophilic | |
Strip Plate | |
High optical quality for reliable results | |
25 Pk., 50 Cases | |
Strip Plate | |
400 μL | |
350 μL | |
For antibody sandwich assays | |
50 | |
Antibodies (Glycoproteins) |
Yes | |
Without Lid | |
Colorimetric | |
Clear | |
Polystyrene | |
96 | |
Round, Flat Bottom (F-well) | |
50 | |
None | |
F8 |