missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Immuno Breakable Modules Clear
Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
£424.00 - £1155.00
Specifications
Format | Strip Plate |
---|---|
Description | High optical quality for reliable results |
Detection Method | Colorimetric |
Color | Clear |
Material | Polystyrene |
Product Code | Brand | Surface Treatment | Well Shape | No. per Pack | Product Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Well Shape | No. per Pack | Product Type | Price | Quantity & Availability | ||
10439962
|
Thermo Scientific™
446442 |
PolySorp | Round, C-bottom | 5 | Strip Plate |
£438.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10506371
|
Thermo Scientific™
446469 |
MaxiSorp | Round, C-bottom | 10 | Strip Plate |
£1155.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10449121
|
Thermo Scientific™
446490 |
MultiSorp | Round, C-bottom | 10 | Frame |
£424.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10450153
|
Thermo Scientific™
446639 |
MaxiSorp | Round | 10 | Microplate Strip |
£443.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10172211
|
Thermo Scientific™
448526 |
MaxiSorp | Round, Starwell | 10 | Microplate Strip |
£437.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10306191
|
Thermo Scientific™
473539 |
PolySorp | Round, C-bottom | 5 | Microplate Strip |
£595.00
Pack of 60 |
Estimated Shipment: 14-11-2024
Log in to see stock. |
||
10459192
|
Thermo Scientific™
473768 |
MaxiSorp | Round, C-bottom | 5 | Strip Plate |
£931.00
Pack of 60 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
10557915
|
Thermo Scientific™
95029390 |
Universal Binding | Round, Flat Bottom (F-well) | 50 | Microplate Strip |
£519.00
Pack of 50 |
In Stock
Log in to see stock. |
||
Description
Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.
- Free-standing modules are easy to handle
Specifications
Strip Plate | |
Colorimetric | |
Polystyrene | |
None |
High optical quality for reliable results | |
Clear | |
96 |